Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBOX5 Mouse anti-Human, Clone: OTI 7G1, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502810

Catalog No. 50-167-3823


Add to cart

Description

Description

UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

UBOX5
Monoclonal
0.87 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O94941
RNF37, UBCE7IP5, UIP5, hUIP5
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22888
Flow Cytometry, Western Blot
OTI 7G1
Unconjugated
UBOX5
Liquid
UBOX5
Human recombit protein fragment corresponding to amino acids 1-130 and 419-487 of human UBOX5 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit