Learn More
UBP 282, Tocris
AMPA/kainate antagonist; distinguishes between motoneuron and dorsal root kainate receptors
Manufacturer:  R&D Systems 1765/50
Quantity 50mg
 View more versions of this product
Catalog No. 17-655-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
333.3
UBP 282
C15H15N3O6
(αS)-α-Amino-3-[(4-carboxyphenyl)methyl]-3, 4-dihydro-2, 4-dioxo-1(2H)-pyrimidinepropanoic acid
Specifications
544697-47-4
>98%
333.3
UBP 282
50mg
C15H15N3O6
(αS)-α-Amino-3-[(4-carboxyphenyl)methyl]-3, 4-dihydro-2, 4-dioxo-1(2H)-pyrimidinepropanoic acid
Safety and Handling
Recommended Storage : Store at Room Temperature