Learn More
Invitrogen™ UBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113574
Description
Immunogen sequence: GASQTSGEQI QPSATIQETQ QWLLKNRFSS YTRLFSNFSG ADLLKLTKED For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Guinea Pig: 92%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 92%.
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.
Specifications
UBP1 | |
Polyclonal | |
Unconjugated | |
UBP1 | |
Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; NF2d9; Nuclear factor 2d9; RGD1564700; transcription factor LBP-1; Ubp1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); upstream-binding protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7342 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZI7 | |
UBP1 | |
Synthetic peptide directed towards the middle region of human UBP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.