Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550696
Description
The antibody detects endogenous levels of total UBP1 protein.
UBP1 Polyclonal antibody specifically detects UBP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
UBP1 | |
Polyclonal | |
Unconjugated | |
UBP1 | |
Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; NF2d9; Nuclear factor 2d9; RGD1564700; transcription factor LBP-1; Ubp1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); upstream-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22221, 7342 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q811S7, Q9NZI7 | |
UBP1 | |
Synthetic peptide corresponding to a region derived from internal residues of human upstream binding protein 1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction