Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBP1112, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
339526-74-8
C10H14NO5P
259.198
LLEOLNUKLKCAFG-UHFFFAOYSA-N
2-amino-2-3-methyl-4-phosphonophenyl propanoic acid
5022489
2-amino-2-(3-methyl-4-phosphonophenyl)propanoic acid
CC1=C(C=CC(=C1)C(C)(C(=O)O)N)P(=O)(O)O
This item is not returnable. View return policy
339526-74-8
C10H14NO5P
259.198
LLEOLNUKLKCAFG-UHFFFAOYSA-N
2-amino-2-3-methyl-4-phosphonophenyl propanoic acid
5022489
2-amino-2-(3-methyl-4-phosphonophenyl)propanoic acid
CC1=C(C=CC(=C1)C(C)(C(=O)O)N)P(=O)(O)O

Group III mGlu antagonist

A selective group III mGlu receptor antagonist (apparent Kd values are 5.1 and 488 μM for group III and group II mGlu receptors respectively; IC50 > 1 mM for group I, NMDA, AMPA and kainate receptors).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 259.2
Percent Purity >99%
Quantity 10mg
Chemical Name or Material UBP1112

Recommended Storage : Store at Room Temperature