Learn More
UBQLN3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233721AP
Catalog No. 50-173-4062
Add to cart
Description
Description
This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis.
Specifications
Specifications
UBQLN3
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C5U9, Q9H347
TUP 1, Ubiquilin 3, UBQLN3
Rabbit
IgG
150 μL
-20°C
Primary
244178, 50613
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBQLN3
Liquid
UBQLN3
UBQLN3 Fusion Protein Ag19982
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse