Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548905

Catalog No. PA548905

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

UBXN10
Polyclonal
PBS with 0.09% sodium azide
Q96LJ8
UBX domain-containing protein 10, UBX domain-containing protein 3, UBXN10, UBXD3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
127733
Western Blot
Unconjugated
UBXN10
Liquid
UBXN10
KLH conjugated synthetic peptide between 157-185 amino acids from the Central region of human UBXN10.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit