Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN2A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548180

Catalog No. PA548180

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

UBXN2A
Polyclonal
PBS with 0.09% sodium azide
P68543, Q99KJ0
UBX domain-containing protein 2A, UBX domain-containing protein 4, UBXN2A, UBXD4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
165324, 217379
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBXN2A
Liquid
UBXN2A
KLH conjugated synthetic peptide between 166-195 amino acids from the C-terminal region of human UBXN2A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit