Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBXN7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561735
Description
Immunogen sequence: TATNHQGLPA VDSEILEMPP EKADGVVEGI DVNGPKAQLM LRYPDGKREQ ITLPEQAKLL ALVKHVQSKG YPNERFELLT NFPRR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
Ubiquitin-binding adapter that links a subset of NEDD8-associated cullin ring ligases (CRLs) to the segregase VCP/p97, to regulate turnover of their ubiquitination substrates.
Specifications
UBXN7 | |
Polyclonal | |
Unconjugated | |
UBXN7 | |
A630090P18; AI196514; KIAA0794; mKIAA0794; UBX domain containing 7; UBX domain protein 7; UBX domain-containing protein 7; Ubxd7; UBXN7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94888 | |
UBXN7 | |
Recombinant protein corresponding to Human UBXN7. Recombinant protein control fragment (Product #RP-100193). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction