Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCB 35625, Tocris Bioscience™

Catalog Number p-4784746
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
202796-42-7
C30H37Cl2IN2O2
655.44
FOAFBMYSXIGAOX-UHFFFAOYSA-N
unii-hd099hot7h, unii-h942hgf14d, hd099hot7h, chembl34626, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , trans, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, trans, unii-e32u2esr7j component foafbmysxigaox-kekkvsgzsa-n, unii-e32u2esr7j component foafbmysxigaox-mumzcososa-n
6918496
1-[(cyclooct-1-en-1-yl)methyl]-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
[I-].CC[N+]1(CC2=CCCCCCC2)CCC(CC1)NC(=O)C1C2=C(OC3=C1C=C(Cl)C=C3)C=CC(Cl)=C2
This item is not returnable. View return policy
202796-42-7
C30H37Cl2IN2O2
655.44
FOAFBMYSXIGAOX-UHFFFAOYSA-N
unii-hd099hot7h, unii-h942hgf14d, hd099hot7h, chembl34626, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , trans, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, trans, unii-e32u2esr7j component foafbmysxigaox-kekkvsgzsa-n, unii-e32u2esr7j component foafbmysxigaox-mumzcososa-n
6918496
1-[(cyclooct-1-en-1-yl)methyl]-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
[I-].CC[N+]1(CC2=CCCCCCC2)CCC(CC1)NC(=O)C1C2=C(OC3=C1C=C(Cl)C=C3)C=CC(Cl)=C2

Potent CCR1 and CCR3 antagonist

Potent chemokine CCR1 and CCR3 receptor antagonist. Inhibits MIP-1α-induced chemotaxis in CCR1 transfectants and eotaxin-induced chemotaxis in CCR3 transfectants (IC50 values are 9.57 and 93.8 nM respectively). Antagonizes CCR3-mediated entry of HIV-1 isolate 89.6 into NP-2 cells (IC50 = 57 nM).

Isomer also available.

Quantity 1 mg
Formula Weight 655.44
Percent Purity >98%
Chemical Name or Material UCB 35625
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit