Learn More
UCHL1 Rabbit anti-Human, Unconjugated, Clone: AAAC-21, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20318
Catalog No. 89-336-718
Add to cart
Description
Description
UCHL1 Monoclonal antibody specifically detects UCHL1 in Human samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF, Flow Cyt.
Specifications
Specifications
UCHL1
AAAC-21
ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
UCHL1
PARK5, PGP9.5, Uch-L1
A synthetic peptide corresponding to human UCHL1
100 μg
Primary
7345