Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UFC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549526

Catalog No. PA549526

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (UFM1; MIM 610553) (Komatsu et al., 2004 [PubMed 15071506]).
Specifications

Specifications

UFC1
Polyclonal
PBS with 0.09% sodium azide
Q9Y3C8, Q9CR09
Ubiquitin-fold modifier-conjugating enzyme 1, Ufm1-conjugating enzyme 1, UFC1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51506, 66155
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UFC1
Liquid
UFC1
KLH conjugated synthetic peptide selected from the C-terminal region of human Ufc1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit