Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UFSP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560315
Description
Immunogen sequence: LAFQLATPNE IFLKKALKHV LSDLSTKLSS NALVFRICHS SVYIWPSSDI NTIPGELTDA SACKNILRFI QFEPEED Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 69%.
Ubiquitin-fold modifier 1 (Ufm1)-specific protease 2 (UfSP2) is a cysteine protease that is responsible for the release of Ufm1 from Ufm1-conjugated cellular proteins, as well as for the generation of mature Ufm1 from its precursor.
Specifications
UFSP2 | |
Polyclonal | |
Unconjugated | |
Ufsp2 | |
1810047C23Rik; BHD; C4orf20; RGD1311161; UFM1 specific peptidase 2; UFM1-specific peptidase 2; ufm1-specific protease 2; UfSP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUQ7 | |
Ufsp2 | |
Recombinant protein corresponding to Human UFSP2. Recombinant protein control fragment (Product #RP-95831). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction