Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGCGL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17930020UL

 View more versions of this product

Catalog No. NBP17930020


Add to cart

Description

Description

UGCGL2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

UGCGL2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
UGGT2
Synthetic peptide directed towards the middle region of human UGCGL2The immunogen for this antibody is UGCGL2. Peptide sequence LCNNPKTKESKLKAAARIVPEWVEYDAEIRQLLDHLENKKQDTILTHDEL.
Immunogen affinity purified
RUO
Primary
55757
Western Blot
Western Blot 1:1000
NP_064506
FLJ10873, HUGT2, MGC117360, UDP-Glc:glycoprotein glucosyltransferase 2, UDP-glucose ceramide glucosyltransferase-like 1, UDP-glucose glycoprotein glucosyltransferase 2
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit