Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGGT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | UGGT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170764
|
Novus Biologicals
NBP170764 |
100 μL |
Each of 1 for $436.00
|
|
Description
UGGT1 Polyclonal specifically detects UGGT1 in Human samples. It is validated for Western Blot.Specifications
UGGT1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ21763, GT, HUGT1, UDP--Glc:glycoprotein glucosyltransferase, UDP-glucose ceramide glucosyltransferase-like 1, UDP-glucose glycoprotein glucosyltransferase 1, UGCGL1, UGGT, UGT1, UGTR | |
UGGT1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
A8KAK1 | |
56886 | |
Synthetic peptides corresponding to UGCGL1 (UDP-glucose glycoprotein glucosyltransferase 1) The peptide sequence was selected from the middle region of UGCGL1. Peptide sequence AAVRIVPEWQDYDQEIKQLQIRFQKEKETGALYKEKTKEPSREGPQKREE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title