Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGGT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA561630

Catalog No. PIPA561630


Add to cart

Description

Description

Immunogen sequence: GEMGIVSNGR FLGPLDEDFY AEDFYLLEKI TFSNLGEKIK GIVENMGINA NNMSDFIMKV DALMSSVPKR ASRYDVTFLR ENHSVIKTNP QENDMFFNV Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 39%.

UDP-glucose:glycoprotein glucosyltransferase (UGT) is a soluble protein of the endoplasmic reticulum (ER) that selectively reglucosylates unfolded glycoproteins, thus providing quality control for protein transport out of the ER.
Specifications

Specifications

UGGT2
Polyclonal
Unconjugated
UGGT2
Liquid
UGGT2
Recombit protein corresponding to Human UGGT2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NYU1
HUGT2, UGCGL2, UGT2, UDP--Glc:glycoprotein glucosyltransferase 2, UDP-Glc:glycoprotein glucosyltransferase 2, UDP-glucose ceramide glucosyltransferase-like 1, UDP-glucose ceramide glucosyltransferase-like 2, UDP-glucose:glycoprotein glucosyltransferase 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55757
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only