Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT1A10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | UGT1A10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023766
|
Novus Biologicals
NBP294621100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023767
|
Novus Biologicals
NBP29462120UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
UGT1A10 Polyclonal antibody specifically detects UGT1A10 in Human samples. It is validated for Western BlotSpecifications
UGT1A10 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
GNT1, UDP glucuronosyltransferase 1 family, polypeptide A10, UDP glycosyltransferase 1 family, polypeptide A10, UDP-glucuronosyltransferase 1-10, UDP-glucuronosyltransferase 1A10, UDP-glucuronosyltransferase 1-J, UDPGT, UDPGT 1-10, UGT1, UGT1*10, UGT1.10, UGT1-10, UGT-1J, UGT1JEC 2.4.1.17 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human UGT1A10 (NP_061948.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS (pH 7.3), 50% glycerol | |
54575 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title