Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT1A9 Mouse anti-Human, Clone: 14G11, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | UGT1A9 |
---|---|
Clone | 14G11 |
Concentration | 1.0 mg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS222925
|
MilliporeSigma
MABS222925UG |
25 μg |
Each for $177.00
|
|
|||||
MABS2229100
|
MilliporeSigma
MABS2229100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
UGT1A9 Monoclonal antibody specifically detects UGT1A9 in Human samples. It is validated for Immunocytochemistry.Specifications
UGT1A9 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
14G11 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
KLH-conjugated linear peptide corresponding to 32 amino acids from the N-terminal half of human UDP-glucuronosyltransferase 1-9 (UDT1A9). | |
Protein G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title