Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT2B11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548590

Catalog No. PA548590


Add to cart

Description

Description

Predicted to react with monkey based on sequence homology.

UDPGT is of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
Specifications

Specifications

UGT2B11
Polyclonal
PBS with 0.09% sodium azide
O75310
UDP-glucuronosyltransferase 2B11, UDPGT 2B11, UGT2B11
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10720
Western Blot
Unconjugated
UGT2B11
Liquid
UGT2B11
KLH conjugated synthetic peptide between 429-458 amino acids from the C-terminal region of human UGT2B11.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit