Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT2B28 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549259

Catalog No. PA549259

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with monkey based on sequence homology.

UDPGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This isozyme has glucuronidating capacity with steroid substrates such as 5-beta-androstane 3-alpha,17-beta- diol, estradiol, ADT, eugenol and bile acids. Only isoform 1 seems to be active.
Specifications

Specifications

UGT2B28
Polyclonal
PBS with 0.09% sodium azide
Q9BY64
UDP-glucuronosyltransferase 2B28, UDPGT 2B28, UGT2B28
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54490
Western Blot
Unconjugated
UGT2B28
Liquid
UGT2B28
KLH conjugated synthetic peptide between 218-246 amino acids from the Central region of human UGT2B28.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit