Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT2B28 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31099725UL

 View more versions of this product

Catalog No. NB120142


Add to cart

Description

Description

UGT2B28 Polyclonal specifically detects UGT2B28 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

UGT2B28
Unconjugated
PBS buffer, 2% sucrose
EC 2.4.1.17, UDP Glucuronosyltransferase 2 Family, Polypeptide B28, UDP Glycosyltransferase 2 Family, Polypeptide B28, UDP-Glucuronosyltransferase 2B28, UDPGT 2B28
The immunogen is a synthetic peptide directed towards the middle region of mouse UGT2B28. Peptide sequence KVLWKFDGKTPATLGHNTRVYKWLPQNDLLGHPKTKAFVTHGGANGVYEA
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
54490
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit