Learn More
UGT2B4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154251AP
Catalog No. 50-173-0219
Add to cart
Description
Description
UGT2B4 belongs to the UDP-glycosyltransferase family. UDPGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This isozyme is active on polyhydroxylated estrogens (such as estriol, 4-hydroxyestrone and 2-hydroxyestriol) and xenobiotics (such as 4-methylumbelliferone, 1-naphthol, 4-nitrophenol, 2-aminophenol, 4-hydroxybiphenyl and menthol). It is capable of 6 alpha-hydroxyglucuronidation of hyodeoxycholic acid.
Specifications
Specifications
UGT2B4
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06133
HLUG25, UDPGT 2B4, UDPGTh 1, UGT2B11, UGT2B4
Rabbit
IgG
150 μL
-20°C
Primary
7363
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UGT2B4
Liquid
UGT2B4
UGT2B4 Fusion Protein Ag8058
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human