Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-UH 232 maleate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
1217473-50-1
C22H33NO5
391.508
MEQAJDYHKYAPJE-GUUGRXDUSA-N
+-uh 232 maleate,cis-+-5-methoxy-1-methyl-2-di-n-propylamino tetralin maleate
46830250
(Z)-but-2-enedioic acid;(1S,2R)-5-methoxy-1-methyl-N,N-dipropyl-1,2,3,4-tetrahydronaphthalen-2-amine
CCCN(CCC)C1CCC2=C(C1C)C=CC=C2OC.C(=CC(=O)O)C(=O)O
1217473-50-1
C22H33NO5
391.508
MEQAJDYHKYAPJE-GUUGRXDUSA-N
+-uh 232 maleate,cis-+-5-methoxy-1-methyl-2-di-n-propylamino tetralin maleate
46830250
(Z)-but-2-enedioic acid;(1S,2R)-5-methoxy-1-methyl-N,N-dipropyl-1,2,3,4-tetrahydronaphthalen-2-amine
CCCN(CCC)C1CCC2=C(C1C)C=CC=C2OC.C(=CC(=O)O)C(=O)O

D2 autoreceptor antagonist. Also D3 partial agonist

D2 antagonist (Ki = 72.7 nM in a ligand binding assay; apparent KB = 14.5 nM in a cAMP accumulation assay); displays preferential activity at central dopamine autoreceptors. Stimulates a marked acceleration of dopamine synthesis and turnover. Produces locomotor stimulation. Exhibits little or no activity at central noradrenalin and 5-HT receptors. Also D3 partial agonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 391.51
Percent Purity >98%
Chemical Name or Material (+)-UH 232 maleate

Recommended Storage : Store at Room Temperature