Learn More
Mac-Mod UltraShield WW Pre-Column Filter, 0.5 um porosity, stainless steel Filter element, Waters port, Waters male, (5/pk)
SDP
Supplier:  Mac-Mod MMUS1505WW
Encompass
Engineered specifically for use in fast, high efficiency UHPLC separations requiring high mobile phase velocities and ultra-high pressures. The ultra-low dispersion of UltraShield Pre-Column Filters maintains the efficiency of UHPLC columns, assuring no loss of critical resolution. With its simple design, UltraShield installs on any analytical, UHPLCcolumn in seconds and is leak tight to 1000 bar (15,000 psi). Simply finger tighten initially, then wrench tighten beyond finger tight an additional quarter turn. Available in 3 unique formats Ultrashield, Ultrashield WW and Ultrashield WP.The UltraShield is compatible with Agilent, Shimadzu, Thermo, Dionex, Hitachi and other popular HPLC instrument brands.The UltraShield WW Pre-Column Filter is compatible with Waters instrumentation and Waters columns.The UltraShield WP Pre-Column Filter is compatible with Waters columns and Non-Waters UHPLC instrumentation.
Catalog No. 50-149-6531