Learn More
YMC America, Inc UHPLC SIZE EXCLUSION COLUMN
SDP
Supplier:  YMC America, Inc DL30S021546PTH
Encompass_Preferred
UHPLC size exclusion column, YMC-Pack Diol 2um. Features: Improvement in resolution and throughput on size exclusion chromatography; more accurate quantitation of monoclonal antibodies with two products; Diol-300 for aggregates analysis.
Catalog No. NC1699896
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.