Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHRF1 Mouse anti-Human, Mouse, Clone: 2B3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA532945

Catalog No. PIMA532945


Add to cart

Description

Description

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

UHRF1
Monoclonal
Unconjugated
UHRF1
Liquid
Uhrf1
Synthetic peptide, corresponding to the N-terminus of Human UHRF1.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
2B3
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8VDF2, Q96T88
Np95; hNP95; ICBP90; RNF106; FLJ21925; MGC138707; E3 ubiquitin-protein ligase UHRF1; Nuclear protein 95; HuNp95; hNp95; RING finger protein 106;
Mouse
IgG2b
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18140, 29128
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit