Learn More
UHRF1 Mouse anti-Human, Unconjugated, Clone: 3A11, Abnova™
Mouse Monoclonal Antibody.
Manufacturer:  Abnova Corporation H00029128M01J
Catalog No. 89-335-119
Add to cart
Description
Description
UHRF1 Monoclonal antibody specifically detects UHRF1 in Human samples. It is validated for WB-Ce, IF, S-ELISA, ELISA, WB-Re.
Specifications
Specifications
UHRF1
3A11
ubiquitin-like with PHD and ring finger domains 1
NM_013282.1
Mouse
IgG1, Kappa
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunofluorescence, Western Blot
Unconjugated
UHRF1
FLJ21925, ICBP90, MGC138707, Np95, RNF106, hNP95
UHRF1 (NP_037414.1, 694 a.a. ∼ 793 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
100 μg
Primary
29128