Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHRF1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549428

Catalog No. PA549428


Add to cart

Description

Description

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Uhrf1
Polyclonal
Unconjugated
Uhrf1
Liquid
UHRF1
KLH conjugated synthetic peptide between 596-628 amino acids from the C-terminal region of Mouse Uhrf1.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8VDF2
E3 ubiquitin-protein ligase UHRF1, 632-, Nuclear protein 95, Nuclear zinc finger protein Np95, Ubiquitin-like PHD and RING finger domain-containing protein 1, mUhrf1, Ubiquitin-like-containing PHD and RING finger domains protein 1, Uhrf1, Np95
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18140
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit