Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHRF1 Mouse anti-Human, Mouse, Clone:, Invitrogen™

Catalog No. PIMA524766
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA524766 100 μL
1 options

Catalog No. PIMA524766

Supplier: Thermo Scientific MA524766

Mouse Monoclonal Antibody

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UHRF1
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 0.1M tris glycine with 150nM NaCl, 50% glycerol and 0.02% sodium azide
Gene UHRF1
Gene Accession No. Q8VDF2, Q96T88
Gene Alias Ac2-121;AL022808;E3 ubiquitin-protein ligase UHRF1;EC 6.3.2.-;FLJ21925;Hnp95 Huhrf1; HuNp95;ICBP90;Inverted CCAAT box binding protein of 90 kDa;Inverted CCAAT box binding protein, 90-Kd;Inverted CCAAT box-binding protein of 90 kDa;Liver regeneration-related protein LRRG126;MGC138707;NP95;Nuclear phosphoprotein, 95-KD;Nuclear protein 95;Nuclear zinc finger protein Np95;RING finger protein 106;RNF106;Transcription factor ICBP90; Ubiquitin like containing PHD and RING finger domains protein 1;Ubiquitin like PHD and RING finger domain containing protein 1;Ubiquitin-like PHD and RING finger domain-containing protein 1;Ubiquitin-like protein containing PHD and RING finger domains 1; Ubiquitin-like with PHD and ring finger domains 1;Ubiquitin-like, containing PHD and RING finger domains, 1;Ubiquitin-like-containing PHD and RING finger domains protein 1;UHRF1;UHRF1_HUMAN.
Gene Symbols UHRF1
Host Species Mouse
Immunogen Human UHRF1 recombit protein
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18140, 29128
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.