Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-1 Antibody (237104), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2588H
Description
ULBP-1 Monoclonal antibody specifically detects ULBP-1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| ULBP-1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Immunology | |
| 80329 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 237104 | |
| Flow Cytometry, CyTOF-ready | |
| ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
| BaF3 mouse pro-B cell line transfected with mouse ULBP-1/MULT-1 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction