Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-2 Rabbit anti-Human, DyLight 650, Clone: 22, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290060C

Catalog No. NB037187


Add to cart

Description

Description

ULBP-2 Monoclonal antibody specifically detects ULBP-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ULBP-2
22
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate
ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Recombinant Human ULBP-2 protein
Protein A purified
RUO
Primary
80328
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit