Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-6/RAET1L Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306628C
This item is not returnable.
View return policy
Description
ULBP-6/RAET1L Polyclonal antibody specifically detects ULBP-6/RAET1L in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
ULBP-6/RAET1L | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
DyLight 650 | |
retinoic acid early transcript 1L protein, retinoic acid early transcript 1L, ULBP6 | |
Produced in rabbits immunized with purified, recombinant Human ULBP-6/RAET1L (AAK91503.1; Met1-Ser217). | |
0.1 mL | |
Immunology | |
154064 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction