Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ULBP2 Monoclonal Antibody (1F12C2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538656
Description
This antibody has been tested in indirect ELISA.
ULBP2 encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells. The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety, or secreted. Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells. This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6.
Specifications
ULBP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9BZM5 | |
ULBP2 | |
Purified recombinant fragment of human ULBP2 (aa: 26-216) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot, Immunocytochemistry | |
1F12C2 | |
Unconjugated | |
ULBP2 | |
ALCAN-alpha; N2DL2; N2DL-2; NKG2D ligand 2; NKG2DL2; RAET1H; Retinoic acid early transcript 1H; UL16 binding protein 2; UL16-binding protein 2; ULBP2; UNQ463/PRO791 | |
Mouse | |
Protein G | |
RUO | |
80328 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction