Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ULK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598600
Description
ULK2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.
Specifications
ULK2 | |
Polyclonal | |
Unconjugated | |
ULK2 | |
A830085I22Rik; ATG1B; AU015340; EC:2.7.11.1; KIAA0623; mKIAA0623; serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; ULK2; unc-51 like autophagy activating kinase 2; unc-51 like kinase 2; Unc51.2; unc-51-like kinase 2 | |
Rabbit | |
Protein G | |
RUO | |
9706 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.61 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8IYT8 | |
ULK2 | |
Recombinant Human Serine/threonine-protein kinase ULK2 protein (211-276AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction