Learn More
Perkin Elmer LLC Ultima Gold Quenched Standards Set - 20ML
Supplier:  Perkin Elmer LLC 6007602
Encompass_Preferred
Ultima Gold quenched standards ensure accurate correction of variable quench when using liquid scintillation cocktails from our Ultima Gold Family of safer scintillation cocktails. These standards are supplied in flame-sealed, low potassium, glass vials. Both conventional size standards and small vial Pico standards are available.
Catalog No. NC1540562
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.