Learn More
High-Tech Conversions ULTIMATE 60™ Polyester Sealed Edge Cleanroom Wipers
High-Tech Conversions ULTIMATE 60™ Polyester Sealed Edge Cleanroom Wipers for Ultra-Critical ISO 3-4 (Class 1-10) Cleanrooms
$759.00
Specifications
Autoclavable No
Basis Weight 115 gsm
Color White
Disposable Yes
Edges Sealed
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
50-135-4706
View Documents
High-Tech Conversions
ULT6098
9 x 8 in.
Case of 5000 for $759.00
Add to cart
50-135-4707
View Documents
High-Tech Conversions
ULT6099/DEL
9 x 9 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
50-135-4708
View Documents
High-Tech Conversions
ULT601212/DEL
12 x 12 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Standard weight
  • Sealed-Edges
  • ISO 3-4 Cleanroom wipes
  • Double bagged
  • Meets USP 797 and USP 800 requirements
Specifications
Specifications
No
White
Sealed
Polyester
Single
Non-sterile
115 gsm
Yes
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
150/Bag
Knit Polyester Cleanroom Wipes