Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Pop Up Pools™
SDP

Instantly expand for quick response to emergency spills

Supplier:  Youngstown Barrel & Drum 8066 UT

 View more versions of this product

Catalog No. 19-130-4812

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Storage bags included with steel models only. Storage bags for economy models also available.

Description

Description

Excellent for response to damaged saddle tanks and cross-over lines, hazardous materials spills, leaking containers, machinery and piping. Can also be used as a decon pool or for collection of used sorbents.

  • Help minimize environmental damage and related cleanup costs
  • Constructed of 10mil thick polyethylene for excellent chemical resistance to diesel fuel, antifreeze, acids, caustics and corrosives
  • Unique foam ring rises with level of liquid to raise sidewalls
  • Compact folded size stores efficiently inside spill kits
  • Sprung-steel pools feature a band of sprung steel to provide rigidity to outer circumference
  • Storage bags available separately (see cat. no. 19-130-4808)

Pools are reusable after proper decontamination.

Order Info

Contact your Fisher Safety Customer Service Representative for ordering information.

Specifications

Specifications

66 gal.
Polyethylene
Used as decontamination pool and collection pool for soiled sorbents
Sprung Steel band
10 mil
249.8 L
Economy Ultra-Pop Up Pool
Storage bags
Chemical
12 x 42 x 53 in. (30.5 x 107 x 135 cm)
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit