Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Pop Up Pools™
SDP

Catalog No. p-3077316
Click to view available options
Capacity (Metric):
75.7 L
567.8 L
Dimensions (L x W x H):
8 x 28 x 35 in.
12 x 64 x 76 in.
Model:
Sprung Steel band

Includes

Storage bags included with steel models only. Storage bags for economy models also available.

Instantly expand for quick response to emergency spills

Excellent for response to damaged saddle tanks and cross-over lines, hazardous materials spills, leaking containers, machinery and piping. Can also be used as a decon pool or for collection of used sorbents.

  • Help minimize environmental damage and related cleanup costs
  • Constructed of 10mil thick polyethylene for excellent chemical resistance to diesel fuel, antifreeze, acids, caustics and corrosives
  • Unique foam ring rises with level of liquid to raise sidewalls
  • Compact folded size stores efficiently inside spill kits
  • Sprung-steel pools feature a band of sprung steel to provide rigidity to outer circumference
  • Storage bags available separately (see cat. no. 19-130-4808)

Pools are reusable after proper decontamination.

Order Info

Contact your Fisher Safety Customer Service Representative for ordering information.

Specifications

Capacity (English) 150 gal.
Capacity (Metric) 567.8 L
Material Polyethylene
Type Economy Ultra-Pop Up Pool
For Use With (Application) Used as decontamination pool and collection pool for soiled sorbents
Includes Storage bags
Model Sprung Steel band
Resistant Chemical
Thickness 10 mil
Dimensions (L x W x H) 12 x 64 x 76 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.