Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Sidewinder Ramp

Ramps are specifically designed to go over the Ultra-Sidewinder Cable Protection System, (Medium). These have a nonskid surface that's good for foot traffic, wheelchairs, mobility scooters or other special needs and golf carts. Light weight enough for easy handling and portability.

$1067.69 - $1590.64

Specifications

Color Black
Type Cable Protection Ramp
For Use With (Equipment) Ultra-Sidewinders (medium)
Material Polymer
For Use With (Application) Cover and protect Sidewinder cable protection system
View More Specs

Products 2
Catalog Number Mfr. No. Weight (Metric) Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Dimensions (L x W x H) Price Quantity & Availability  
19-339-058
View Documents Promotion Details
Youngstown Barrel & Drum
1822 UT
8.5 kg 48 x 36 x 3 in.
Each for $1,067.69
Add to cart
 
19-339-059
View Documents Promotion Details
Youngstown Barrel & Drum
1824 UT
17 kg 70 x 37 x 4 in.
Each for $1,590.64
Add to cart
 
Description

Description

Nonskid surface, good for foot traffic, wheelchairs, mobility scooters or other special needs and golf carts. Light weight enough for easy handling and portability.
Specifications

Specifications

Black
Ultra-Sidewinders (medium)
Cover and protect Sidewinder cable protection system
Cable Protection Ramp
Polymer
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit