Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Pallets, Steel

Provides secondary containment for chemicals and hazardous materials. 100% leak-free tested. Excellent chemical resistance - will not melt or deform from extreme heat or fire. Forkliftable for easy handling and transport. Removable grating for easy clean-up and maintenance.

$709.37 - $1075.00

Specifications

Capacity (Metric) 257 L
Model Steel
For Use With (Application) Catch material from leaking drums
Includes Grate
Color Steel
View More Specs

Products 2
Catalog Number Mfr. No. Type Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Type Dimensions (L x W x H) Price Quantity & Availability  
19-171-400
View Documents Promotion Details
Youngstown Barrel & Drum
1180 UT
Spill Pallet 47.25 x 31.5 x 17.875 in.
Each for $709.37
Add to cart
 
19-171-401
View Documents Promotion Details
Youngstown Barrel & Drum
1182 UT
Spill Pallet 47.25 x 47.25 x 13.25 in.
Each for $1,075.00
Add to cart
 
Description

Description

Secondary containment for chemicals and hazardous materials. Will not melt or deform from extreme heat or fire.
Specifications

Specifications

257 L
Catch material from leaking drums
Steel
Chemical
Steel
Grate
Meets SPCC and EPA Container Storage Regulation 40 CFR 264.175 Spill Containment Regulations.
Spill Pallet
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit