Learn More
Andwin ScientificSupplier Diversity Partner ULTRA THERMOMETER -50/250C DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 23609174
These Long-Stem Thermometers allow the measuring of temperature even in hard-to-reach areas, such as laboratory glasswares. These thermometers also have an LCD screen that shows temperature in either unit (�C or �F).
Catalog No. 50-112-6023