Learn More
Thermo Scientific™ Ultrac™ Adapters
For use with Centrifuges from Thermo Scientific
Products 1
Catalog Number Mfr. No. Tube Type Price Quantity & Availability  
Catalog Number Mfr. No. Tube Type Price Quantity & Availability  
05-375-9
View Documents
Thermo Scientific™
072260F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Adapters for models Centra™-CL3/CL3R, -GP8/GP8R; HN-SII; EXD and K; and discontinued Centra-MP4/MP4R. Note: rotors require 05-375-60 Shaft Adapter when used on Centra-CL3/-CL3R.
  • With stainless-steel handles for easy loading
  • Autoclavable plastic construction provides full support along length of tube
  • Numbered tube slots for easy identification
  • Withstand centrifugal forces to 50,000xG