Learn More
Beaver Visitec Intl Ltd Cellulose Eye Spear 100/SP
Supplier:  Beaver Visitec Intl Ltd 40410
Encompass
Made from highly absorbent; natural; cellulose material. Fast wicking action delivers quick absorption of blood and fluid from surgical area. The flat tip provides precise application at the surgical incision.
Catalog No. NC1651161
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.