Thermo Scientific™ UltraFast™ UFC-1 GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
Manufacturer: Thermo Scientific™ UFMC00001010401
 View more versions of this product
Catalog No. 03-170-387
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Trace GC UltraFast Instrument
    Stationary Phase UltraFast UFC-1
    Item Description UFMC00001010401
    Diameter 0.1mm
    Film Thickness 0.1µm
    Length (Metric) 5m
    Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
    Applications:
    Chemical
    Petrochemical
    Environmental
    Flavors and fragrances
    Tailored Specifically for Robustness and Speed
    UltraFast GC columns utilize short (2–10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
    Dramatically shorter analysis times
    Increased sample throughput–by a factor of 20
    Longer column lifetimes
    UltraFast UFC-1 Uses:
    ASTM D-2887
    General
    ISO 9377-2
    ASTM D-3710
  • Description & Specifications
Related Products
Product Content Feedback