Learn More
Thermo Scientific™ UltraFast™ UFC-WAX GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
Manufacturer:  Thermo Scientific™ UFMC00001010501
 View more versions of this product
Catalog No. 03-170-388
Add to cart
Description
Description
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
Chemical
Petrochemical
Environmental
Flavors and fragrances
Tailored Specifically for Robustness and Speed
UltraFast GC columns utilize short (2 to 10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
Dramatically shorter analysis times
Increased sample throughput–by a factor of 20
Longer column lifetimes
UltraFast UFC-WAX Columns uses:
Fatty acid methyl esters (FAMEs)
Essential oils
General
Specifications
Specifications
0.1 µm
Ultrafast
5 m x 0.1 mm x 0.1 µm
5 m
GC for Chemicals, GC for Environmental Analysis, GC for Flavors and Fragrances, GC for Petrochemicals
UltraFast UFC-WAX
Trace GC UltraFast Instrument
0.1 mm