Learn More
Thermo Scientific™ UltraFast™ UFC-WAX GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
$2830.00
Specifications
Length (Metric) 5 m
Stationary Phase UltraFast UFC-WAX
View More Specs
Products 2
Catalog Number Mfr. No. Film Thickness Price Quantity & Availability  
Catalog Number Mfr. No. Film Thickness Price Quantity & Availability  
03-170-388
View Documents
Thermo Scientific™
UFMC00001010501
0.1 µm Each for $2,830.00
Add to cart
03-170-389
View Documents
Thermo Scientific™
UFMC00001010503
0.2 µm Each for $2,830.00
Add to cart
Description
Description
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
Chemical
Petrochemical
Environmental
Flavors and fragrances
Tailored Specifically for Robustness and Speed
UltraFast GC columns utilize short (2 to 10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
Dramatically shorter analysis times
Increased sample throughput–by a factor of 20
Longer column lifetimes
UltraFast UFC-WAX Columns uses:
Fatty acid methyl esters (FAMEs)
Essential oils
General
Specifications
Specifications
5 m
UltraFast UFC-WAX