Learn More
Epredia™ UltraFit™ Nylon Glove Liners
Nylon glove liners are ideal for those who have sensitivities to latex, nitrile, and vinyl materials. A form fitting design and seamless fingertips ensure optimum dexterity and tactile sensitivity.
$38.00
Specifications
Color White
Disposable No
Glove Cuff Style Knit
Hand Ambidextrous
Inspected for Medical Use No
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
14-379-45 Edge
View Documents
Epredia™
5100312001
Large
Pack of 12 for $38.00
Case of 12 PK for $455.00
Edge
Add to cart
14-379-35
View Documents
Epredia™
5100112001
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
51-002-12001
View Documents
Epredia™
5100212001
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Inspected for laboratory and cleanroom use
  • Helps prevent irritation caused by latex, nylon, and vinyl gloves
  • Non-linting, ultra-thin nylon construction
  • Seamless fingertips
  • Form-fitting design
  • Washable
Specifications
Specifications
White
Knit
No
Nylon
No
Ambidextrous
Yes