Learn More
Thermo Scientific™ UltraFIT™ Nylon Glove Liners Available on GSA/VA Contract for Federal Government customers only.
Protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving.
$66.80 - $70.80
Specifications
Type Nylon Glove Liners
View More Specs
Products 5
Catalog Number Mfr. No. Size Includes Laboratory/Cleanroom Use Price Quantity & Availability  
Catalog Number Mfr. No. Size Includes Laboratory/Cleanroom Use Price Quantity & Availability  
14-379-35 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5100112001
Small 12 Pr./Pk. Yes Pack of 12 for $66.80
Case of 12 PK for $669.00
Add to cart
14-379-38 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5100212001I
Medium 12 Pr./Pk. No Pack of 12 for $70.80
Case of 12 PK for $725.50
Add to cart
14-379-45 Edge Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5100312001
Large 12 Pr./Pk. Yes Pack of 12 for $66.80
Case of 12 PK for $669.00
Edge
Add to cart
14-379-39 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5100312001I
Large 12 Pr./Pk. No Pack of 12 for $70.80
Case of 12 PK for $725.50
Add to cart
51-002-12001
View Documents
Thermo Scientific™
5100212001
Medium 12 Pr./Pk. Yes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving with Thermo Scientific™ UltraFIT™ Nylon Glove Liners. These liners are are pre-shrunk for multiple laundering.
  • Form-fitting design for a skin-tight fit
  • Ultra-thin fabric for optimum dexterity and sense of touch
  • Extra layer of insulation when working in cooler areas
  • Seamless fingertips
  • Cuffs are color coded for easy sorting
  • Nonlinting and washable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Nylon Glove Liners