Learn More
MilliporeSigma™ Ultrafree™-MC Centrifugal Filters
Ideal centrifugal filter for protein extraction, protein purification and sample prep. Ultrafree-MC Centrifugal Filter is a single-use, disposable filter.
$128.50
Products 1
Catalog Number Mfr. No. Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Price Quantity & Availability  
UFC30LG25
View Documents
MilliporeSigma™
UFC30LG25
0.2 μm
Each for $128.50
Add to cart
Description
Description
Ideal centrifugal filter for protein extraction, protein purification and sample prep. Ultrafree-MC Centrifugal Filter is a single-use, disposable filter.
Ultrafree™-MC (0.5 mL) and Ultrafree™-CL (2 mL) centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes (from 0.1 to 5.0 µm) for fast filtration and highly reproducible performance. Pre-sterilized units are also available. Use in fixed-angle rotors for 1.5 mL tubes (-MC) and 15 mL tubes (–CL).
  • Five different pore sizes from 0.1 to 5.0 µm
  • Pre-sterilized units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 mL tubes
Applications
  • Removal of particles and precipitates from aqueous and some solvent-based samples
  • Ideal for use with protein and nucleic acid solutions