Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ultramarine Blue, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
57455-37-5
57455-37-5

UL105, 57455-37-5

Spectrum™ Ultramarine Blue
CAS Max % 1
Grade Reagent
Packaging Fiber Drum
Quantity 25 kg